missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ≈95% N-Hexane, for HPLC

Product Code. 10499170 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10499170 1 L Amber Glass Bottle 1 L
10101910 2.5 L Plastic-coated Glass Bottle 2.5 L
10234150 5 L Aluminum Can 5 L
10703611 2.5 L Amber Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10499170 Supplier Fisher Chemical Supplier No. H/0406/15
  • ONLINE EXCLUSIVE PRICE
    159.40 €
    136.00 € / 1 L
    Save 23.40 €
    14% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis.

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 160mbar at 20°C
Viscosity 0.31 mPaS at 20°C
Boiling Point 69°C
Molecular Weight (g/mol) 86.18
Formula Weight 86.18g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexanes, 95% n-Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.