missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 99%, pure, mixture of isomers

Product Code. 10267563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10267563 2.5 L Glass Bottle 2.5 L
10677201 1 L EcoSafPak™ Glass Bottle 1 L
10073443 10 L Plastic Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10267563 Supplier Thermo Scientific Acros Supplier No. 411540025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
Melting Point -95.0°C
Name Note Mixture of Isomers
CAS 92112-69-1, 110-54-3
Color Colorless
Density 0.6590g/mL
Quantity 2.5 L
Flash Point −22°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Assay Percent Range 99%
Specific Gravity 0.659
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Vapor Pressure Vapor Pressure: 160mbar at 20°C
MDL Number MFCD00009520
Viscosity 0.31 mPa.s (20°C)
Beilstein 01, 142
Merck Index 14, 4694
Synonym Hex
Boiling Point 69.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Percent Purity 99%
Grade Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexanes
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hexane, branched andlinear

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.