missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, ACS, 98.5+%

Product Code. 11377648
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
4 x 1 L
Unit Size:
1 L
2.5 L
4 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377648 2.5 L 2.5 L
11405000 500 mL 500mL
11367648 1 L 1 L
11347648 4 x 1 L 4 L
11357648 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11377648 Supplier Thermo Scientific Alfa Aesar Supplier No. 033321.K4
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009520
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.67 g/mL
Quantity 2.5 L
Flash Point <-23°C (-9°F)
Physical Form Liquid, Mixture Of 5 Isomers
Refractive Index 1.3795
UN Number UN1208
Boiling Point 65°C to 70°C
Solubility Information Miscible with alcohol,chloroform,acetone and ether. Immiscible with water.
Formula Weight 86.18
Percent Purity ≥98.5%
Grade ACS
Chemical Name or Material Hexanes, Mixed isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.