missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, Optima™ for HPLC and GC, Fisher Chemical™

Product Code. 15537059 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15537059 1 L Wide Neck Amber Bottle 1 L
15547059 4 L Amber Soda Lime Glass 4 L
15557059 4 L Safe-Cote™ Amber Glass  4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15537059 Supplier Fisher Chemical Supplier No. H3031
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

High purity and versatile solvent suitable for HPLC, spectrophotometry, GC/MS, and pesticide residue analysis | CAS: 92112-69-1 | C6H14 | 86.18 g/mol

High purity and versatile solvent suitable for HPLC, spectrophotometry, GC/MS, and pesticide residue analysis

  • Mixture of hexanes where n-hexane content is ≥60%
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis and tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Absorbance 1 AU max. at 195nm, 0.25 AU max. at 210nm, 0.07 AU max. at 220nm, 0.005 AU max. at 254nm
Color Colorless
Density 0.659g/cm³
Quantity 1 L
Flash Point −95°C (−139°F)
Identification Pass test
Packaging Wide Neck Amber Bottle
Physical Form Liquid
Refractive Index 1.3740 to 1.3780
Residue after Evaporation 1ppm max.
Assay Percent Range 99.9% min by GC
Vapor Pressure 160mbar at 20°C
Viscosity 0.31 mPa/s at 20°C
UN Number 1208
Water 0.01% Max.
Boiling Point 69°C
Formula Weight 86.18
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Hexanes
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexanes
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: hexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.