missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, technical, mixture of isomers

Product Code. 13228003 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
HDPE Bottle
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13228003 5 L HDPE Bottle 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 13228003 Supplier Thermo Scientific Acros Supplier No. 444340050
  • ONLINE EXCLUSIVE PRICE
    111.00 €
    101.00 € / 5 L
    Save 10.00 €
    9% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
CAS 92112-69-1, 110-54-3
Melting Point -95.0°C
Density 0.6590g/mL
Boiling Point 69.0°C
Flash Point −22°C
Packaging HDPE Bottle
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Refractive Index 1.3748 to 1.3810
Quantity 5 L
Specific Gravity 0.659
Synonym Hex
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Molecular Weight (g/mol) 86.18
Viscosity 0.31 mPa.s (20°C)
Formula Weight 86.18
Grade Technical
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.