missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Hexanios G + R DOSES

Product Code. 11720425
Change view
Click to view available options
Quantity:
200 Pk.
50 Pk.
Unit Size:
1.25 L
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11720425 200 Pk. Pack of 200
11745625 50 Pk. 1.25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11720425 Supplier Laboratoires Anios™ Supplier No. 165097ZD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleaning and pre-disinfection of surgical and medical instrumentation, medical devices, thermosensitive instrumentation and endoscopic equipment. Cleaning in ultrasonic bins. Collecting of stained instrumentation.

Pre-disinfectant detergent of instrumentation :

  • Clear blue solution.
  • Couple HEXANIOS G+R / STERANIOS 2% or ANIOXYDE 1000.
    for the manual reprocessing of endoscopes validated
    (BIOTECH GERMANDE).
  • Non corrosive with instrumentation (norme NF S 94 402-1).
  • Usable in ultra-sonic bath.
  • pH of diluted producte : approx 7.
  • Physico-chemical stability up to +70°C.

Specifications

Product Type Hexanios G + R
Form Liquid
Container Type Pouch
Quantity 200 Pk.
Volume (Metric) 25 mL
compliance-icons compliance-icons
Product Identifier
  • HEXANIOS G+R
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: didecyldiammonium chloride, tetrasodiumEDTA, propan-2-ol, lauryldimethylamine oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.