missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Hexanios™ G + R Pre-disinfectant Detergent

Product Code. 11822663
Change view
Click to view available options
Volume (Metric):
25 mL
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11822663 25 mL 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11822663 Supplier Laboratoires Anios™ Supplier No. 165046

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleaning and pre-disinfection of surgical and medical instrumentation, medical devices, thermosensitive instrumentation and endoscopic equipment. Cleaning in ultrasonic bins. Collecting of stained instrumentation.

Pre-disinfectant detergent of instrumentation:

  • Clear blue solution.
  • Couple HEXANIOS G+R / STERANIOS 2% or ANIOXYDE 1000.
    for the manual reprocessing of endoscopes validated
    (BIOTECH GERMANDE).
  • Non corrosive with instrumentation (norme NF S 94 402-1).
  • Usable in ultra-sonic bath.
  • pH of diluted producte : approx 7.
  • Physico-chemical stability up to +70°C.

Specifications

Form Liquid
pH 7
Appearance Blue
Certifications/Compliance EN 1040, NF T 72-171, 72-190 and NF T T72–300 / 301, BK
Container Type Bottle
Content And Storage 5°C to 35°C
Volume (Metric) 25 mL
compliance-icons compliance-icons
Product Identifier
  • HEXANIOS G+R
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501-Dispose of contents/container to .
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: PROPYLENE GLYCOL, DIDECYLDIMETHYLAMMONIUM CHLORIDE, TETRASODIUM ETHYLENE DIAMINETETRAACETATE, PROPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.