missing translation for 'onlineSavingsMsg'
Learn More

Hexanophenone, 98%

Product Code. 11457736
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457736 100 g 100g
11437736 5 g 5g
11447736 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457736 Supplier Thermo Scientific Alfa Aesar Supplier No. A14712.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 942-92-7 | C12H16O | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942-92-7
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009512
InChI Key MAHPVQDVMLWUAG-UHFFFAOYSA-N
Synonym hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone
PubChem CID 70337
IUPAC Name 1-phenylhexan-1-one
SMILES CCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 25°C to 27°C
Density 0.95
Boiling Point 265°C
Flash Point >110°C (230°F)
Refractive Index 1.5105
Quantity 100 g
Beilstein 1908667
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material Hexanophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.