missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl chloride, 97%

Product Code. 10723361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723361 100 g Glass bottle 100g
10363432 5 g Glass bottle 5g
10733361 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10723361 Supplier Thermo Scientific Acros Supplier No. 169041000
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 142-61-0 | C6H11ClO | 134.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-61-0
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.61
MDL Number MFCD00000760
InChI Key YWGHUJQYGPDNKT-UHFFFAOYSA-N
Synonym caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
PubChem CID 67340
IUPAC Name hexanoyl chloride
SMILES CCCCCC(=O)Cl

Specifications

Melting Point -87.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 151.0°C to 153.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4COCl
Refractive Index 1.4253 to 1.4273
Quantity 100 g
Beilstein 02, 324
Merck Index 15, 1765
Specific Gravity 0.95
Solubility Information Solubility in water: hydrolysis. Other solubilities: decomposes in alcohol, soluble in ether, chloroform, ethylether and, acetone
Formula Weight 134.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Hexanoyl chloride, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexanoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.