missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl Glycine-13D2,15N, TRC

Product Code. 30404573
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404573 10 mg 10mg
30404210 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404573 Supplier Toronto Research Chemicals Supplier No. TRCH28120210MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanoyl Glycine-13C2,15N
Molecular Formula C613C2H1515NO3
Quantity 10 mg
Synonym N-(1-Oxohexyl)glycine-13C2,15N, Caproylglycine-13C2,15N, N-Caproylglycine-13C2,15N, NSC 224460, n-Hexanoylglycine-13C2,15N
InChI Formula InChI=1S/C8H15NO3/c1-2-3-4-5-7(10)9-6-8(11)12/h2-6H2,1H3,(H,9,10)(H,11,12)/i6+1,8+1,9+1
SMILES O=C([15N][13C][13C](O)=O)CCCCC
Molecular Weight (g/mol) 176.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.