missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Hexanoyl-L-carnitine-d3 HCl (N-methyl-d3) |
| CAS | 2483831-95-2 |
| Molecular Formula | C13 D3 H23 N O4 . Cl |
| Quantity | 0.05 g |
| Synonym | Hexanoyl-L-carnitine-d3 HCl (N-methyl-d3), 1-Propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxohexyl)oxy]-, chloride (1:1), (2 R)- (ACI), Hexanoyl-L-carnitine-d3 Chloride, Hexanoyl-L-carnitine-D3 (N-methyl-D3) chloride, (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxohexoxy)-1-propanaminium chloride, [(2 R)-3-Carboxy-2-(hexanoyloxy)propyl]dimethyl(methyl-d3)ammonium chloride |
| InChI Formula | InChI=1 S/C13H25NO4.ClH/c1-5-6-7-8-13(17)18-11(9-12(15)16)10-14(2,3)4;/h11H,5-10H2,1-4H3;1 H/t11-;/m1./s1/i2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCC |
| IUPAC Name | [(2 R)-3-carboxy-2-hexanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 298.8213 |
| Formula Weight | 298.1739 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction