missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl-L-carnitine-d3 HCl (N-methyl-d3), TRC

Product Code. 30398431
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398431 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398431 Supplier Toronto Research Chemicals Supplier No. TRCH29444150MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanoyl-L-carnitine-d3 HCl (N-methyl-d3)
CAS 2483831-95-2
Molecular Formula C13 D3 H23 N O4 . Cl
Quantity 50 mg
Synonym Hexanoyl-L-carnitine-d3 HCl (N-methyl-d3), 1-Propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxohexyl)oxy]-, chloride (1:1), (2R)- (ACI), Hexanoyl-L-carnitine-d3 Chloride, Hexanoyl-L-carnitine-D3 (N-methyl-D3) chloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxohexoxy)-1-propanaminium chloride, [(2R)-3-Carboxy-2-(hexanoyloxy)propyl]dimethyl(methyl-d3)ammonium chloride
InChI Formula InChI=1S/C13H25NO4.ClH/c1-5-6-7-8-13(17)18-11(9-12(15)16)10-14(2,3)4;/h11H,5-10H2,1-4H3;1H/t11-;/m1./s1/i2D3;
SMILES [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCC
IUPAC Name [(2R)-3-carboxy-2-hexanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molecular Weight (g/mol) 298.8213
Formula Weight 298.1739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.