Learn More
Thermo Scientific Chemicals Hexokinase, yeast
Used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.
151.00 € - 304.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15415969
|
Thermo Scientific Chemicals
J63795.4I |
1 kU |
151.00 €
1kg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15425969
|
Thermo Scientific Chemicals
J63795.J+ |
10 kU |
304.00 €
10kg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Hexokinase is used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.