missing translation for 'onlineSavingsMsg'
Learn More

Hexylamine, TRC

Product Code. 30391148
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391148 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391148 Supplier Toronto Research Chemicals Supplier No. TRCH295665100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexylamine
CAS 111-26-2
Molecular Formula C6 H15 N
Quantity 100 mL
Synonym 1-Hexanamine (9CI, ACI), Hexylamine (6CI, 8CI), 1-Aminohexane, 1-Hexaneamine, 1-Hexylamine, ENT 16554, Mono-n-hexylamine, n-Hexylamine, NSC 2590
InChI Formula InChI=1S/C6H15N/c1-2-3-4-5-6-7/h2-7H2,1H3
SMILES CCCCCCN
IUPAC Name hexan-1-amine
Molecular Weight (g/mol) 101.19
Formula Weight 101.1204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.