missing translation for 'onlineSavingsMsg'
Learn More

Hexylmagnesium chloride, 2M solution in THF, AcroSeal™

Product Code. 11975941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975941 800 mL AcroSeal™ Glass Bottle 800mL
11965941 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975941 Supplier Thermo Scientific Acros Supplier No. 434688000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 44767-62-6 | C6H13ClMg | 144.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44767-62-6 , 109-99-9
Molecular Formula C6H13ClMg
Molecular Weight (g/mol) 144.93
MDL Number MFCD06798087
InChI Key ODOOOEXDBNGOOF-UHFFFAOYSA-M
Synonym hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10877247
SMILES CCCCCC[Mg]Cl

Specifications

Boiling Point 65.0°C to 68.0°C
Color Brown to Brown-Gray
Physical Form Solution
Quantity 800 mL
Flash Point −25°C
Solubility Information Solubility in water: reacts
IUPAC Name magnesium;hexane;chloride
Formula Weight 144.93
Concentration or Composition (by Analyte or Components) 1.90 to 2.20M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.965
Density 0.9650g/mL
Chemical Name or Material Hexylmagnesium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexylmagnesium chloride, 2M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.