Learn More
LNI Swissgas HG PRO 900
Product Code. 17812309 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage Unit Size
17812309 100/115/220/230 V Each
1 options
Product Code. 17812309 Supplier LNI Swissgas Supplier No. 6920.15.090
  • ONLINE EXCLUSIVE PRICE
    13585.00 €
    11783.53 € / Pack of 1
    Save 1801.47 €
    13% Off
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Hydrogen generator ultra high purity.
  • Accessory
Specifications
Voltage 100/115/220/230 V
Type Hydrogen Carrier Gas Generator