missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HIC (Hydrophobic Interaction Chromatography) Media

Product Code. 11638873 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Capto Butyl media
Capto Phenyl (High Sub) media
Ligand Type:
Butyl
Phenyl
Unit Size:
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Ligand Type Unit Size
11638873 Capto Butyl media Butyl 25mL
11344063 Capto Phenyl (High Sub) media Phenyl 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11638873 Supplier Cytiva Supplier No. 17545901
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Features high chemical stability, properties to increase productivity and epoxy coupling

  • High flow rate, high capacity and low backpressure reduce cycle times
  • Chemically stable to commonly used aqueous buffers
  • Epoxy chemical coupling

  • Capto Phenyl (high sub) has a hydrophobic ligand of phenyl and is a hydrophobic interaction chromatography medium used for protein purification
  • Capto Butyl has a hydrophobic ligand of butly and a highly cross-linked agarose matrix

Used in capture and intermediate stages of protein purification

TRUSTED_SUSTAINABILITY

Spécification

Product Type Capto Butyl media
Flow Rate 600 cm/Hoμr
For Use With (Application) For Used in The Capture and Intermediate Stages of Protein PurifiCatIon
Quantity 25 mL
Ligand Type Butyl
Media Type Highly Cross-Linked Agarose
Max. Pressure 300 kPa
Particle Size 75 μm
pH Range 3 to 13
Storage Requirements 4°C to 30°C
Storage Buffer 20% Ethanol
Binding Capacity 27 mg BSA/mL
Height (Metric) Bed 20 cm
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Capto Butyl, 25 ml
Mention d'avertissement
  • Warning
Catégorie de danger
  • Flammable liquid Category 3
Mention de danger
  • H226-Flammable liquid and vapour.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informations supplémentaires
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.