missing translation for 'onlineSavingsMsg'
Learn More

Higenamine Hydrochloride, TRC

Product Code. 30134193
Change view
Click to view available options
Quantity:
50 mg
250 mg
5 g
Unit Size:
250mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134193 5 g 5g
30003313 250 mg 250mg
30004402 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134193 Supplier Toronto Research Chemicals Supplier No. TRCH3258185G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11041-94-4
Molecular Formula C16 H17 N O3 . Cl H
Molecular Weight (g/mol) 307.77
Synonym 1-(4-Hydroxybenzyl)-1,2,3,4-tetrahydroisoquinoline-6,7-diol Hydrochloride
IUPAC Name 1-[(4-hydroxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrochloride
SMILES Cl.Oc1ccc(CC2NCCc3cc(O)c(O)cc23)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H17NO3.ClH/c18-12-3-1-10(2-4-12)7-14-13-9-16(20)15(19)8-11(13)5-6-17-14;/h1-4,8-9,14,17-20H,5-7H2;1H
Formula Weight 307.0975
Percent Purity >95%
Chemical Name or Material Higenamine Hydrochloride
Grade HPLC
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.