missing translation for 'onlineSavingsMsg'
Learn More

Chemgene™ High Level Surface Disinfectant

Product Code. 15166686
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15166686 1 L 1 L
15176686 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15166686 Supplier Chemgene™ Supplier No. XTM304
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Combines enhanced active ingredient molecules with micelle cleaning technology to provide swift penetration of the cell walls to ensure rapid cell death. Chemgene™ High Level Surface Disinfectant ensures that cell death is achieved rather than just suspending activity.

  • Biodegradable
  • Non-corrosive
  • Long shelf life
  • Diluted concentrate lasts up to 6 months
  • Compliant with REACH regulations and the Biocidal Product directive

Specifications

Quantity 6/Pk.
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • CHEMGENE HLD4H CONCENTRATE
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P315-Get immediate medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.