Learn More
Abeba™ High Original 5009 Shoes
Product Code. 15621778
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. 15621778
Brand: Abeba™ 5009/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe includes PU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • Black
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
  • Narrow fit
  • Wedge heel 45mm
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color Black
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 5009