missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ High-Power USB Wi-Fi Module

Product Code. 15521973 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 module
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15521973 1 module Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15521973 Supplier Applied Biosystems™ Supplier No. A26774
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-Power USB Wi-Fi Module

Upgrade your wireless-enabled Applied Biosystems™ instrument with this High-Power USB Wi-Fi Module for high speed transfer rates and maximum range of connectivity during remote monitoring of your instrument.

Key features and benefits include:
  • Encrypted data security standards
  • Leading wireless technology standard for top performance and range
  • Maximum speed (up to 300 Mbps) for fast wireless transfer rates
  • Maximum range with built-in high-power antenna

    Note: This module is not suitable for the QuantStudio™ 3D Digital PCR Instrument. A different Wi-Fi module is offered for this instrument (A28598).
  • TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Equipment) End-point Thermal Cycler
    Product Line Thermal Cycler Accessories
    Product Type USB Wi-Fi Module
    For Use With (Application) Real Time PCR (qPCR)
    Shipping Condition Room Temperature
    Quantity 1 module

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.