missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ High-Purity Cyclohexyl Maltoside Detergents

Product Code. 30245344 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Reagent Type:
Cymal-5
Cymal-6
Cymal-7
Unit Size:
1g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Reagent Type Unit Size
30245344 1 g Cymal-5 1g
30245087 5 g Cymal-5 5g
30245136 1 g Cymal-6 1g
30245146 5 g Cymal-6 5g
30245343 1 g Cymal-7 1g
30245216 5 g Cymal-7 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 30245344 Supplier Thermo Scientific™ Supplier No. A65517
  • ONLINE EXCLUSIVE PRICE
    230.00 €
    200.10 € / 1g
    Save 29.90 €
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Cyclohexyl maltoside detergents are a class of specialized non-ionic detergents designed for the solubilization, stabilization, and manipulation of biomolecules, particularly membrane proteins.

Cyclohexyl maltoside detergents are a class of specialized non-ionic detergents designed for the solubilization, stabilization, and manipulation of biomolecules, particularly membrane proteins. They provide efficient solubilization of membrane proteins from lipid bilayers, allowing for their extraction and study in solution. Cyclohexyl maltoside detergents are widely used in protein purification, structural biology studies, and other research involving membrane proteins. They can be employed in various experimental techniques, including crystallization, spectroscopy, and functional analysis.

Features of cyclohexyl maltoside detergents
• Gentle—preserves the native conformation and activity of membrane proteins
• Enhanced stability during extraction and downstream applications
• High-purity with low UV absorptivity

Overall, cyclohexyl maltoside detergents are versatile and can be used in various experimental techniques, including protein purification, crystallization, spectroscopy, and structural biology studies. They are compatible with a wide range of biomolecules and can be employed for both soluble and membrane protein research.

5-Cyclohexyl-1-Pentyl-β-D-Maltoside
5-Cyclohexyl-1-Pentyl-β-D-Maltoside (Cymal-5) has a specific combination of a cyclohexyl and pentyl alkyl chain attached to the maltose moiety. This combination of alkyl chains can enhance the detergent's hydrophobicity, solubilization properties, and interactions with certain types of membrane proteins. Cymal-5 is often utilized for solubilization and stabilization of membrane protein complexes. The solubilization efficiency allows for the extraction of intact complexes, helping to preserve their native structure and functionality. It is important to note that the specific solubilization efficiency of Cymal-5 can vary depending on the specific protein, its structural features, and the experimental conditions.

Properties of Cymal-5
CMC: (H2O) ~ 2.4-5.0 mM (0.12%)
Aggregation number: (H2O) ~ 47
Purity: ≥ 99% β+α (by HPLC analysis)
pH: 5-8 (1% solution in water)

6-Cyclohexyl-1-Hexyl-β-D-Maltoside
6-Cyclohexyl-1-Hexyl-β-D-Maltoside (Cymal-6) has a hexyl (C6) alkyl chain attached to the maltose moiety. The longer hexyl alkyl chain in Cymal-6 provides it with greater hydrophobicity compared to other cyclohexyl maltoside detergents. Due to its enhanced hydrophobicity, Cymal-6 may be particularly effective in solubilizing membrane proteins that have higher hydrophobic regions or multiple transmembrane domains that often require stronger detergent interactions. The longer alkyl chain may also lead to the formation of larger micelles. Cymal-6  is well-suited for structural biology studies, such as protein crystallization or nuclear magnetic resonance (NMR) spectroscopy.

Properties of Cymal-6
CMC: (H2O) ~ 0.56 mM (0.028%)
Aggregation number: (H2O) ~ 91
Purity: ≥ 99% β+α (by HPLC analysis)
pH: 5-8 (1% solution in water)

7-Cyclohexyl-1-Heptyl-β-D-Maltoside
7-Cyclohexyl-1-Heptyl-β-D-Maltoside (Cymal-7) has a specific combination of a cyclohexyl and heptyl alkyl chain attached to the maltose moiety. The presence of both cyclohexyl and heptyl alkyl chains in Cymal-7 contributes to its hydrophobicity. Its specific alkyl chain combination can contribute to improved stability, helping to maintain the structural integrity and functionality of proteins during solubilization and downstream applications. The longer alkyl chain may also lead to the formation of larger micelles. Cymal-7 is well-suited for structural biology studies, such as protein crystallization or nuclear magnetic resonance (NMR) spectroscopy.

Properties of Cymal-7
CMC: (H2O) ~ 0.19 mM (0.0099%)
Aggregation number: (H2O) ~ 150
Purity: ≥ 99% β+α (by HPLC analysis)
pH: 5-8 (1% solution in water)

TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Detergent
CAS 250692-65-0
Molecular Formula C23H42O11
Solubility ≥20% (in water at 20°C)
Content And Storage Store below -20°C.
Form Lyophilized
Reagent Type Cymal-5
Quantity 1 g
Molecular Weight (g/mol) 494.5

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.