missing translation for 'onlineSavingsMsg'
Learn More

Cytiva High Purity rNTP Solutions

Product Code. 10784737 Shop All Cytiva Products
Change view
Click to view available options
Format:
Liquid
Product Type:
ATP, 100 mM solution
CTP, 100 mM solution
GTP, 100 mM solution
UTP, 100 mM solution
Quantity:
25 μmol
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Product Type Quantity Unit Size
10784737 Liquid GTP, 100 mM solution 25 μmol Each
10136184 Liquid CTP, 100 mM solution 25 μmol Each
10004164 Liquid UTP, 100 mM solution 25 μmol Each
10676505 Liquid ATP, 100 mM solution 25 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10784737 Supplier Cytiva Supplier No. 27207601
  • ONLINE EXCLUSIVE PRICE
    97.06 €
    84.25 € / Pack of 1
    Save 12.81 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for in vitro transcription or for reactions requiring rNTPs of high purity. Cytiva Lifescience™ High Purity rNTP Solutions provide easy-to-use, time-saving solutions.

  • Each lot is tested for ribonuclease contamination to ensure success in your application
  • High purity: >98% triphosphate

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Description Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
pH 7.5
Product Type GTP, 100 mM solution
Concentration 100 mM
Purity ≥98% triphosphate
Quantity 25 μmol
Color Colorless
For Use With (Application) in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.