Learn More
Applied Biosystems™ High Resolution Melt (HRM) Plug-in for DA3 Software
Shop All Applied Biosystems ProductsSpecifications
Specifications
| For Use With (Equipment) | QuantStudio 1, QuantStudio 3 and 5, QuantStudio 6 and 7 Pro, StepOne and StepOnePlus Real Time PCR Systems, and The 7500 Fast Real Time PCR System |
| Software Compatibility | Diomni Design and Analysis v3.1 Desktop Application, and DA2 Software |
| Quantity | 10 Licenses |
Frequently Asked Questions (FAQs)
You can download the compatible HRM version and the software from our Connect Platform. Select "Design and Analysis 2," and you will also find the High Resolution Melt plugin there. Alternately, you can also visit the following webpage for further information and redirection.
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.