missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ High Resolution Melt Software v3.2

Product Code. 15541533 Shop All Applied Biosystems Products
Change view
Click to view available options
Content And Storage:
Includes One License Key for 1 User
License Key for 10 Users
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
15541533 Includes One License Key for 1 User Each
15531533 License Key for 10 Users Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15541533 Supplier Applied Biosystems™ Supplier No. a30150
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High Resolution Melt Software v3.2 is an all-new and improved tool for High Resolution Melting (HRM) analysis. HRM analysis is a post-PCR analysis method used to identify variation in nucleic acid sequences.

The method is based on detecting small differences in PCR melting (dissociation) curves. It is enabled by high-brightness, dsDNA-binding dyes used in conjunction with real-time PCR instrumentation that has precise temperature ramp control, advanced data capture capabilities, and access to software designed specifically for HRM analysis. High Resolution Melt Software v3.2 brings HRM to the QuantStudio 1, QuantStudio 3 and 5, StepOne and StepOnePlus Real-Time PCR Systems, and the 7500 Fast Real-Time PCR System, and contains features that make fast and accurate HRM results more accessible than ever.

Key product features:

• Minimal user input to achieve results—software automatically applies default/saved analysis settings upon opening an experiment file
• Easy recall—new Analysis Settings Library saves settings for future experiments
• Greater accuracy—clustering algorithm accurately distinguishes control and variant genotypes
• Increased sensitivity—software allows input of expected number of clusters for difficult-to-detect SNPs (class 4)
• Ability to achieve more in one run—separate analysis of multiple assays run on a single plate
• Shortcuts in experiment setup—direct copy and paste functions with Excel; well information and plate layout clearly displayed in Plate View

Minimize user input to achieve results and easily recall analysis settings
High Resolution Melt Software v3.2 automatically applies default or saved analysis settings upon opening an experiment file, which allows the user to achieve results with minimal input. It also includes a new Analysis Settings Library, allowing the user to save optimized settings and easily recall them for use in future experiments.

Realize greater accuracy and increase your sensitivity
The v3.2 software utilizes an improved clustering algorithm that helps to accurately distinguish control and variant genotypes, and allows the user to input the expected number of clusters for increased sensitivity for difficult SNP genotyping.

Achieve more in one run
With the v3.2 software, you now have the ability to conduct separate analyses of multiple assays run on a single plate.

Get your experiments up and running fast with shortcuts in setup
Users of High Resolution Melt Software v3.2 can copy and paste directly from Excel spreadsheets to set up experiments quickly. Plate View also clearly displays well information and the plate layout to ease plate setup.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes One License Key for 1 User
For Use With (Equipment) 7500 Fast System, QuantStudio™ 1, QuantStudio™ 3, QuantStudio™ 5, StepOnePlus™ System, StepOne™ System
Shipping Condition Room Temperature
Quantity 1 license

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.