Learn More
Nabertherm High Temperature Furnace LHT 04/17
Product Code. 31041700 Shop All Nabertherm Products
Click to view available options
Includes:
Furnace Only
Gas Connection
Gas Connection, Over-Temp Limiter
Over-Temperature Limiter
Product Code. 31041700
Brand: Nabertherm LHT437WN1
An all-purpose high-temperature furnace ideal for laboratory processes and the sintering of advanced ceramic materials.
  • Tmax 1600°C, 1750°C, or 1800°C
  • High-quality heating elements made of molybdenum disilicide offer very good protection against chemical interaction between charge and heating elements
  • Adjustable air inlet opening
  • Exhaust air opening in the roof
  • Controller with touch operation P580 (50 programs with each 40 segments)
  • Available with:
    • Over-temperature limiter with adjustable cutout temperature as temperature limiter to protect the furnace and load
    • Port for thermocouple in the furnace roof
    • Protective gas connection to purge with non-flammable process, not gas tight
    • Manual or automatic gas supply system
    • Stackable saggars for loading in up to two or three levels, depending on model
Specifications
Controller Type P580
Capacity (Metric) 4 L
Temperature (Metric) 1750°C
Type Muffle Furnace with Lift Door
Includes Furnace Only
Voltage 220/240 V