missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ High Visibility Jacket 8845-Orange

Product Code. 17219871 Shop All Uvex Products
Change view
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17219871 4X-Large Each
17229871 5X-Large Each
17239871 6X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17219871 Supplier Uvex™ Supplier No. 9861515
  • ONLINE EXCLUSIVE PRICE
    121.00 €
    108.76 € / Pack of 1
    Save 12.24 €
    10% Off
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All that is essential is that everyone can be seen despite the poor visibility. The high-visibility clothing from the uvex protection flash collection uses bright reflective strips. And of course the cargo trousers and softshell jackets with their optimum fit can also be seen in daylight

  • Hi-vis warning
  • Weatherproof parka
  • Color: hi-vis orange / navy
  • Material: face fabric: 100% polyester with polyurethane coating, lining and wadding: 100% polyester
  • This functionally warning-weatherproof parka with reflectors and thermo wadding is ideal for the cool and cold season with rain and snow.
  • Highly lockable collar, hood is able to rolling in
  • Reflective stripes on body and sleeves (according to EN 471)
  • Concealed 2-way-in front zip fastener with chin protection facing
  • 2 side pockets, 2 chest pockets, mobile phone pocket, pencil case pocket and inside pocket
  • Contrasting colored inserts on shoulder and sideward front lot
  • Hem and waist width-adjustable
  • Wadded, firm worked into lining with thermo insulation with service-zip fastener to add on emblems
  • Certified according to: EN ISO 20471 X3 EN 343 (3/1)
  • Sizes: S(46/48) - XXL(62/64)
  • Care instruction:30°C washable (gentle wash), no chlorinate, tumbler impossible, ironing impossible, dry-cleaning impossible (max. 25 wash cycles)
TRUSTED_SUSTAINABILITY

Specifications

Size 4X-Large
Gender Men's
Material Polyester
Color Orange
Closure Type Zipper
Disposable Reusable
Product Type Hi-vis Jacket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.