missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ HILED Lighting (GM) Upgrade
GREENER_CHOICE

Product Code. 16327565 Shop All Syngene Products
Click to view available options
Color:
Far Red
Red, Green, Blue and UV
UV
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16327565

Brand: Syngene™ GMHILEDUPGDUV

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

HILED lighting upgrade from RGB or RIR, designed to allow for the addition of UV, Far Red, Red, Green, or Blue.

  • Required equipment: GM-HILED-RGB or GM-HILED-RIR
  • Bag of parts to be fitted by professional
  • Works with GBOX mini only

Specifications

Color UV
For Use With (Equipment) GM-HILED-RGB or GM-HILED-RIR
Product Type Lighting
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.