missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ HILED Lighting Option
GREENER_CHOICE

Product Code. 17172496 Shop All Syngene Products
Click to view available options
Color:
Green, Blue, Far Red
Green, Blue, Infa Red, Far Red
Infa Red, Far Red
Red, Blue, Green
Red, Blue, Green, InfraRed
Red, InfraRed
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17172496

Brand: Syngene™ GMHILEDIRFR

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

HILED lighting option for use with GBOX Mini includes gantry, electronics, and lighting units.

  • For use with GBOX Mini only
  • Including gantry, electronics, and lighting units

Specifications

Color Infa Red, Far Red
For Use With (Equipment) GBOX Mini
Includes Gantry, electronics, lighting units
Product Type Lighting
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.