missing translation for 'onlineSavingsMsg'
Learn More

Hippuric acid, MedChemExpress

Product Code. 16402698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402698 Supplier Medchem Express Supplier No. HYW016562
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hippuric Acid (2-Benzamidoacetic acid), an acyl glycine produced by the conjugation of benzoic acid and glycine, is a normal component in urine as a metabolite of aromatic compounds from food.

Hippuric Acid (2-Benzamidoacetic acid), an acyl glycine produced by the conjugation of benzoic acid and glycine, is a normal component in urine as a metabolite of aromatic compounds from food.

Specifications

Chemical Name or Material Hippuric acid
CAS 495-69-2
Color White
Purity Grade Notes Research
Molecular Formula C9H9NO3
Synonym 2-Benzamidoacetic acid
Solubility Information DMSO : 75 mg/mL (418.60 mM; Need ultrasonic)
SMILES O=C(O)CNC(C1=CC=CC=C1)=O
Molecular Weight (g/mol) 179.17
Quantity 5 g
Formula Weight 179.17
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.