missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Q HP 16/10 Column

Product Code. 13538800 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13538800 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13538800 Supplier Cytiva Supplier No. 29018182
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide fast, preparative separations of proteins and other biomolecules

  • Strong anion exchanger designed for preparative ion exchange chromatography separations
  • Highly cross-linked agarose provides excellent chemical and physical stability
  • Slim, modern design allows for easy handling and connection

TRUSTED_SUSTAINABILITY

Specifications

Description HiPrep Q HP 16/10
Diameter (English) Inner Column 0.6 in.
Flow Rate 1 to 5 mL/min.
For Use With (Application) HiPrep Q HP is a 20 mL strong anion exchange chromatography column for high resolution small-scale protein purification.
Content And Storage 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 20 mL
Quantity 20 mL
compliance-icons
Product Identifier
  • HiPrep Q HP 16/10
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.