missing translation for 'onlineSavingsMsg'
Learn More

Cytiva His Buffer Kit

Product Code. 10074384 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10074384 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10074384 Supplier Cytiva Supplier No. 11003400
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

8X phosphate stock solution (pH 7.4), 2 x 100mL; 2M imidizole ( pH 7.4), 100mL

Purify histidine-tagged proteins under native conditions. His Buffer Kit consists of convenient, premade buffer stock solutions optimized for histidine-tagged protein purification.

  • High-quality buffers with minimal absorbance at 280nm
  • Optimized protocols for His GraviTrap, His SpinTrap, His MultiTrap HP, His MultiTrap FF and HisTrap columns

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material His Buffer Kit
Quantity Each
Includes 2 x 100mL Phosphate 8x stock solution (pH 7.4), 100mL highly pure 2 M Imidazole (pH 7.4), instructions for use
compliance-icons compliance-icons compliance-icons
Product Identifier
  • His Buffer Kit
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Reproductive toxicity Category 1B
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H226-Flammable liquid and vapour.
  • EUH208-Contains nickel. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: imidazole
  • UNKNOWN AQUATIC-Contains 15% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.