missing translation for 'onlineSavingsMsg'
Learn More

Cytiva His GraviTrap™ Columns

Product Code. 11564865 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11564865 10 x 1 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564865 Supplier Cytiva Supplier No. 11003399

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for purification of histidine-tagged proteins using gravity flow. His GraviTrap™ Columns enable fast and simple purification.

  • Prepacked with Ni Sepharose 6 Fast Flow media and ready to use
  • Allows direct purification of both clarified and unclarified cell lysates
  • Protein binding capacity approximately 40mg histidine-tagged protein/column
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.59 in.
Diameter (Metric) Inner 15 mm
Description His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification
Material Polypropylene
pH Range 3 to 12
Support Type 6% Cross-linked Agarose
Content And Storage 4°C to 30°C
Particle Size 45 to 165 μm
Quantity 10 x 1 mL
Type Gravity Flow Column
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains nickel. May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.