missing translation for 'onlineSavingsMsg'
Learn More

Cytiva His SpinTrap™ TALON™

Product Code. 12654506 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 x 100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12654506 50 x 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12654506 Supplier Cytiva Supplier No. 29000593
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Single-use spin column for high purity purification of histidine-tagged proteins

Offers different selectivity compared to nickel-charged chromatography media

Specifications:

  • Ligand - TALON Superflow Cobalt-IMAC
  • Matrix - Cross-linked agarose, 6%
  • Particle size -     60 -160μm
  • Binding Capacity/ml - Up to 20 mg histidine-tagged protein
  • pH stability working range - 3 to 12
  • Complete packsize - 50 x 100μL
  • Bed volume - 100μL

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 x 100 μL
For Use With (Application) For High Purity PurifiCatIon of Histidine-Tagged Proteins
Storage Requirements 4°C to 8°C
Binding Capacity Up to 20 mg Histidine-tagged Protein
Product Type His SpinTrap Talon prepacked Column
Particle Size 60 to 160 μm
Matrix 6% Cross-Linked Agarose
Bed Volume (Metric) 100 μL
Ligand Type Talon Superflow Cobalt-IMAC
compliance-icons
Product Identifier
  • his spintrap talon columns
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.