missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScale™ Columns Packed with Superdex™ Increase or Superose™ Increase Resins

Similar to the new-generation Superdex™ Increase and Superose™ Increase Size Exclusion Chromatography (SEC) Columns, but for use with purifying larger volumes of up to 8.5 mL

Brand:  Cytiva 29321905


Additional Details : CAS Number : 64-17-5 Weight : 1.80000kg


 View more versions of this product

Product Code. 15928872

  • 6683.00 € / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket

Description

Description

  • High-resolution SEC columns
  • Small particle size and narrow particle size distribution of the packed resin ensures highly pure protein
Specifications

Specifications

Size Exclusion Chromatography (SEC)
1 Ea.
3.2 mL
Superdex 200 Increase
2 MPa
Store at 4°C to 30°C, 20% Ethanol
16 x 400 mm
Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Superdex 200 Increase HiScale 16/40

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers