missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen Capto™ L Columns

Product Code. 12661206 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4.7 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12661206 4.7 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12661206 Supplier Cytiva Supplier No. 17547814
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

HiScreen Capto L columns are prepacked with Capto L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.

HiScreen Capto L columns are part of the process development platform available from Cytiva.

  • Prepacked, ready-to-use columns for convenient process development optimization.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume (4.7ml) gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity.

TRUSTED_SUSTAINABILITY

Specifications

Flow Rate <500 cm/hr.
Material Polypropylene
pH Range 2 to 10
Support Type Rigid, highly cross-linked Agarose
Content And Storage 2°C to 8°C
Particle Size 85 μm
Quantity 4.7 mL
For Use With (Application) Optimization and parameter screening
Type Agarose
Ligand Type Recombinant Protein L (E. coli), mammalian free
compliance-icons
Product Identifier
  • hiscreen capto L
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.