missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20 cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography resins using the same linear fluid velocity
Specifications
Specifications
| For Use With (Application) | Hydrophobic Interaction Chromatography |
| Diameter (Metric) Inner | 7.7 mm |
| Height (Metric) Bed | 100 mm |
| Bed Volume (Metric) | 4.7 mL |
| Packaging Quantity | 1 x 4.7 mL |
Safety and Handling
Signal Word
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?