missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Capto™ S ImpAct Prepacked Column
Shop All Cytiva Products
Click to view available options
Quantity:
1 x 4.7 mL
Unit Size:
Each
Description
- 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
- Small bed volume gives fast results and minimal sample/buffer consumption.
- Easily connected in series to achieve 20 cm bed height.
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.
- Prepacked with Capto S ImpAct, a strong cation exchange BioProcess medium, designed for immediate purification and polishing of a wide range of biomolecules, especially monoclonal antibodies.
Specifications
Specifications
| Product Type | HiScreen Capto S ImpAct Prepacked Column |
| Quantity | 1 x 4.7 mL |
| Flow Rate | 2.3 mL/min |
| Particle Size | 90μm |
| Column Type | Ion Exchange Column |
Safety and Handling
Product Identifier
- HiScreen Capto S ImpAct
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction