missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen MabSelect PrismA Column

Product Code. 15893965 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 4.7 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15893965 1 x 4.7 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15893965 Supplier Cytiva Supplier No. 17549815
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein A Column

This column is prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 4.7 mL
Flow Rate <500 cm/hr.
Max. Operating Pressure 3 bar (0.3 MPa, 44 psi)
Quantity 1 x 4.7 mL
For Use With (Application) Antibody affinity chromatography
Type Pre-packed Column
Binding Capacity ≈80 mg Human IgG/mL Resin at 6 min residence time. ≈40 mg Human IgG/mL at 2 min residence time.
compliance-icons
Product Identifier
  • HiScreen MabSelect PrismA column
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P502-Refer to manufacturer/supplier for information on recovery/recycling.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE- per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.