Learn More
Thermo Scientific™ HisPur™ Cobalt Purification Kit, 1 mL
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.
Additional Information and Online Ordering
Highlights:
High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions
Low metal leaching – no metal contamination in eluted histidine-tagged protein sample
Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives
Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates
Cost effective – resin can be reused several times or discarded after single use
Superior – performs better than other commercially available IMAC Resins
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
- Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin
Includes:
Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer
Specifications
Specifications
| Capacity (Metric) | 5 columns |
| Description | HisPur Cobalt Spin Purification Kit, 1mL |
| Content And Storage | Sufficient For: Purifying ∼10 mg of His-tagged protein per column • HisPur Cobalt Spin Columns, 1 mL, 5 columns • Equilibration/Wash Buffer, 100 mL • Elution Buffer, 50 mL Store at 4°C. |
| Product Line | HisPur |
| Quantity | 5 columns |
| Target | His-Tagged Protein |
| Type | Protein Purification System |
| Stationary Phase | Metal Chelate, Divalent Cobalt |
| Sufficient For | 10 mg of His-Tagged Protein per Column |
| Format | Spin Column |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.