missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Spin Purification Kit, 1 mL

Product Code. 11812114 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 Column Kit
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 11812114

Brand: Thermo Scientific™ 88228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.

Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.

The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.

Highlights:

  • High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin

  • Versatile – purify proteins using native or denaturing conditions

  • Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives

  • Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges

  • Cost-effective – reuse the same batch of resin at least five times

  • Easy to use – pre-formulated buffers available for kit formats

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin

Includes:

Kits contain HisPur Ni-NTA Columns

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 mL/Column
Description HisPur Ni-NTA Spin Purification Kit, 1mL
Content And Storage Sufficient For: Purifying up to 60 mg of His-tagged protein per column
• HisPur Ni-NTA Spin Columns, 1 mL, 5 columns
• Phosphate-buffered Saline (10X), 30 mL
• Imidazole (2M), 8 mL
• Column Plugs, 5 plugs

Store at 4°C.
Product Line HisPur
Quantity 5 Column Kit
Target His-Tagged Protein
Type Spin Column Kit
Stationary Phase Divalent Nickel, Metal Chelate
Sufficient For 60 mg of His-Tagged Protein per Column
Format Spin Column
Column Type Affinity, Agarose Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.