missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Histamine dihydrochloride

Product Code. 18792464
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792464 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18792464 Supplier R&D Systems™ Supplier No. 3545/50
  • ONLINE EXCLUSIVE PRICE
    129.94 €
    116.55 € / 50mg
    Save 13.39 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous histamine receptor agonist

Endogenous agonist at histamine receptors (H1-4). Released from mast cells and basophils and exhibits inflammatory, vasodilatory and bronchoconstrictory activity. Stimulates gastric acid secretion and acts as a neurotransmitter in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-92-8
Quantity 50 mg
Product Line Tocris
Molecular Formula C5H11Cl2N3
Formula Weight 184.07
Purity >98%
Molecular Weight (g/mol) 184.06
Chemical Name or Material Histamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.