missing translation for 'onlineSavingsMsg'
Learn More

Histamine Dihydrochloride, TRC

Product Code. 17970855
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970855 5 g 5g
17952325 1 g 1g
17912335 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970855 Supplier Toronto Research Chemicals Supplier No. TRCH4365005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-92-8
Molecular Formula C5 H9 N3 . 2 Cl H
Molecular Weight (g/mol) 184.07
Synonym Histamine dihydrochloride, 2-(1H-imidazol-4-yl)ethanamine dihydrochloride
IUPAC Name 2-(1H-imidazol-4-yl)ethanamine;dihydrochloride
SMILES Cl.Cl.NCCc1c[nH]cn1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H9N3.2ClH/c6-2-1-5-3-7-4-8-5;;/h3-4H,1-2,6H2,(H,7,8);2*1H
Formula Weight 183.033
Percent Purity >95%
Chemical Name or Material Histamine Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.