missing translation for 'onlineSavingsMsg'
Learn More

Histamine, MedChemExpress

Product Code. 16488676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488676 Supplier Medchem Express Supplier No. HYB1204
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Histamine is an organic nitrogenous compound involved in local immune responses as well as regulating physiological function in the gut and acting as a neurotransmitter.

Histamine is an organic nitrogenous compound involved in local immune responses as well as regulating physiological function in the gut and acting as a neurotransmitter.

Specifications

Chemical Name or Material Histamine
CAS 51-45-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C5H9N3
Quantity 100 mg
Synonym Ergamine
Solubility Information H2O : ≥ 34 mg/mL (305.89 mM)
SMILES NCCC1=CN=CN1
Molecular Weight (g/mol) 111.15
Formula Weight 111.15
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.