missing translation for 'onlineSavingsMsg'
Learn More

Histamine

Product Code. 15434269
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15434269 Supplier Thermo Scientific Alfa Aesar Supplier No. J61727.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 51-45-6 | C5H9N3 | 111.15 g/mol

Histamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.

Solubility
Soluble in water, alcohol, and hot chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Hygroscopic. Handle and store under inert gas. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-45-6
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD00005210,MFCD00128939
InChI Key NTYJJOPFIAHURM-UHFFFAOYSA-N
Synonym histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
PubChem CID 774
ChEBI CHEBI:18295
SMILES NCCC1=CN=CN1

Specifications

Color White
Quantity 1 g
UN Number UN2811
Beilstein 2012
Merck Index 14,4719
Solubility Information Soluble in water,alcohol,and hot chloroform.
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine
Formula Weight 111.15
Physical Form Powder
Chemical Name or Material Histamine
compliance-icons compliance-icons
Product Identifier
  • Histamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Imidazole-4-ethanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.