missing translation for 'onlineSavingsMsg'
Learn More

Histamine, TRC

Product Code. 30041306
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041306 250 mg 250mg
30040775 1 g 1g
30041146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041306 Supplier Toronto Research Chemicals Supplier No. TRCH436503250MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-45-6
Molecular Formula C5 H9 N3
Molecular Weight (g/mol) 111.15
Synonym 1H-Imidazole-5-ethanamine, 1H-Imidazole-4-ethanamine (9CI), Histamine (8CI), 2-(1H-Imidazol-4-yl)ethanamine, 2-(1H-Imidazol-4-yl)ethylamine, 2-(1H-Imidazol-5-yl)ethanamine, 2-(1H-Imidazol-5-yl)ethylamine, 2-(3H-Imidazol-4-yl)ethylamine, 2-(4-Imidazolyl)ethanamine, 2-(4-Imidazolyl)ethylamine, 4-(2-Aminoethyl)imidazole, 5-Imidazoleethylamine, Eramin, Ergamine, Ergotidine, Imidazole-4-ethylamine, NSC 33792, β-Imidazolyl-4-ethylamine
IUPAC Name 2-(1H-imidazol-4-yl)ethanamine
SMILES NCCc1c[nH]cn1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H9N3/c6-2-1-5-3-7-4-8-5/h3-4H,1-2,6H2,(H,7,8)
Formula Weight 111.0796
Chemical Name or Material Histamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.