missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Histone Acetyltransferase Inhibitor IV, CPTH2

Catalog Number p-7082482
Click to view available options
Quantity:
5 mg
10 mg
This item is not returnable. View return policy
357649-93-5
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor
This item is not returnable. View return policy
357649-93-5
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor

Histone acetylation plays an important role in regulating the chromatin structure and is tightly regulated by two classes of enzyme, histone acetyltransferases (HAT) and histone deacetylases (HDAC). Deregulated HAT and HDAC activity plays a role in the development of a range of cancers. Consequently, inhibitors of these enzymes have potential as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
TRUSTED_SUSTAINABILITY
Quantity 10 mg
Description Brown
Content And Storage 2°C to 8°C
For Use With (Application) A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
Formula Weight 291.80g/mol
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'