missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz HistoPlast Paraffin Wax

Product Code. 30158393
Change view
Click to view available options
Melting Point:
52°C to 54°C
54°C to 56°C.
56°C to 58°C
58°C to 60°C
60°C to 62°C
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Melting Point Unit Size
30158393 58°C to 60°C Pack of 10
30158519 56°C to 58°C Pack of 10
30158402 60°C to 62°C Pack of 10
30158487 54°C to 56°C. Pack of 10
30158453 52°C to 54°C Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30158393 Supplier Heinz Herenz Supplier No. 1131236
  • ONLINE EXCLUSIVE PRICE
    117.60 €
    83.90 € / Pack of 10
    Save 33.70 €
    28% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HistoPlast paraffin wax, supplied in pearl shape. For tissue section infiltration and embedding.

  • HistoPlast paraffin wax
  • Supplied in pearl shape
  • For tissue section infiltration and embedding
  • Compound of purified paraffin and plastic polymers, without dimethyl sulfoxide (DMSO).
  • Cutting thickness 2μm.

Specifications

Melting Point 58°C to 60°C
Product Type Paraffin
Form Pellet
compliance-icons compliance-icons
Product Identifier
  • X10 HistoPlast Paraffin Wax, supplied in pearl sha pe, melting point 58-60 degrees
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Paraffin waxes and Hydrocarbon waxes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.