missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HistoScreen™ Cassettes in Tube Packs

Product Code. 16343936 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16343936 Beige Pack of 750
16363936 Green Pack of 750
16373936 Gray Pack of 750
16383936 Lavender Pack of 750
16393936 Light Green Pack of 750
16303946 Lilac Pack of 750
16313946 Orange Pack of 750
16323946 Pink Pack of 750
16333946 Tan Pack of 750
16343946 White Pack of 750
16353946 Yellow Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 16343936 Supplier Epredia™ Supplier No. B851001BE
  • ONLINE EXCLUSIVE PRICE
    452.00 €
    350.70 € / Pack of 750
    Save 101.30 €
    22% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Eliminates the need for sponges or bags, ideal for very small biopsy tissue samples. Cassettes are compatible with Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in 12 color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY

Specifications

Pore Style Mesh
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
Color Beige
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type HistoScreen Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.