missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX HitHunter™ cGMP Assay
Click to view available options
Quantity:
10,000 dp (384-well)
Unit Size:
1 set
Description
The HitHunter™ cGMP Assay is a detection kit offering a robust, sensitive and easy-to-use assay to measure intracellular cGMP levels. This product uses EFC technology, and contains ED-tagged cGMP, anti-cGMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cGMP, the anti-cGMP antibody will be sequestered by binding to this untagged cGMP, allowing for EFC between the tagged cGMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit may be used with Eurofins DiscoverX cell lines as well as cell lines from other sources. This detection kit contains reagents for 25 plates (384 well).
Specifications
Specifications
| Assay Entry | GPCR |
| For Use With (Application) | Detection Kits |
| Quantity | 10,000 dp (384-well) |
| Target Entry | cGMP |
| Target | cGMP |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction